missing translation for 'onlineSavingsMsg'
Learn More

Ethanesulfonyl chloride, 98+%

Catalog No. AAB2372336
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2372336 500 g
AAB2372314 25 g
AAB2372322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2372336 Supplier Thermo Scientific Chemicals Supplier No. B2372336
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 594-44-5 | C2H5ClO2S | 128.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 594-44-5
Molecular Formula C2H5ClO2S
Molecular Weight (g/mol) 128.57
MDL Number MFCD00007460
InChI Key FRYHCSODNHYDPU-UHFFFAOYSA-N
Synonym ethylsulfonyl chloride, ethylsulfochloride, 1-ethanesulfonyl chloride, ethanesulphonyl chloride, ethanesulfonylchloride, unii-b56zoc898a, ethylsulfonylchloride, ethansulfonyl chloride, ethansulfonylchlorid, ethanesulfonylchoride
PubChem CID 11667
IUPAC Name ethanesulfonyl chloride
SMILES CCS(Cl)(=O)=O

Specifications

Melting Point -70°C
Density 1.35
Boiling Point 176°C to 178°C
Flash Point 83°C (181°F)
Refractive Index 1.454
Linear Formula CH3CH2SO2Cl
Quantity 500 g
UN Number UN2927
Beilstein 773865
Sensitivity Moisture sensitive
Formula Weight 128.57
Percent Purity ≥98%
Chemical Name or Material Ethanesulfonyl chloride
Show More Show Less
Hazard Category H227-H302-H314-H331
Hazard Statement GHS H Statement
H301-H310-H330-H314-H318-H227
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber 209-842-5
RTECSNumber KI8050000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only