missing translation for 'onlineSavingsMsg'
Learn More

Ethambutol dihydrochloride

Catalog No. AAJ6069514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6069514 25 g
AAJ6069506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6069514 Supplier Thermo Scientific Chemicals Supplier No. J6069514

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An antibacterial agent | CAS: 1070-11-7 | C10H26Cl2N2O2 | 277.23 g/mol

Ethambutol dihydrochloride it acts as a tuberculostatic antibacterial agent. It is used to treat tuberculosis in combination with other antibiotics to reduce the development of drug resistance. It has been used to study multidrug-resistant tuberculosis (MDR-TB) in highest-incidence-rate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 1070-11-7
Molecular Formula C10H26Cl2N2O2
Molecular Weight (g/mol) 277.23
MDL Number MFCD00216025
InChI Key AUAHHJJRFHRVPV-HSTMFJOWSA-N
Synonym ethambutol 2hcl, 2r-2-2-2r-1-hydroxybutan-2-yl amino ethyl amino butan-1-ol dihydrochloride, ethambutol related compound b usp, prestwick_208, aronis24117, 2,2'-1,2-ethanediyldiimino bis-1-butanol dihydrochloride, unii-jng307dj5x component, 2,2'-1,2-ethanediyldi imino di 1-butanol dihydrochloride, 2r-2-2-2r-1-hydroxybutan-2-yl amino ethylamino butan-1-ol dihydrochloride, 2r,2'r-2,2'-ethane-1,2-diylbis azanediyl bis butan-1-ol dihydrochloride
PubChem CID 6419929
IUPAC Name (2R)-2-[2-[[(2R)-1-hydroxybutan-2-yl]amino]ethylamino]butan-1-ol;dihydrochloride
SMILES CCC(CO)NCCNC(CC)CO.Cl.Cl

Specifications

Color White
Quantity 25 g
Merck Index 14,3720
Solubility Information Slightly soluble in water
Formula Weight 277.23
Physical Form Powder
Chemical Name or Material Ethambutol dihydrochloride
Hazard Category H360
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 213-970-7
RTECSNumber EL3854000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only