missing translation for 'onlineSavingsMsg'
Learn More

Estriol, 97%, Thermo Scientific Chemicals

Catalog No. p-7077841
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460490010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC460490010 Supplier Thermo Scientific Chemicals Supplier No. 460490010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50-27-1
Molecular Formula C18H24O3
Molecular Weight (g/mol) 288.38
InChI Key PROQIPRRNZUXQM-ZXXIGWHRSA-N
Synonym estriol, oestriol, estratriol, ovestin, ovestrion, trihydroxyestrin, destriol, tridestrin, aacifemine, oestratriol
PubChem CID 5756
ChEBI CHEBI:27974
IUPAC Name (8R,9S,13S,14S,16R,17R)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,16,17-triol
SMILES CC12CCC3C(C1CC(C2O)O)CCC4=C3C=CC(=C4)O

Specifications

Melting Point 280.0°C to 282.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 288.38
Percent Purity 97%
Chemical Name or Material Estriol
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Suspected of causing cancer.
May damage fertility or the unborn child.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
EINECSNumber 200-022-2

RUO – Research Use Only