missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Estramustine, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 2998-57-4 |
| Quantity | 25 mg |
| Linear Formula | C23H31Cl2NO3 |
| Formula Weight | 440.40 |
| Grade | BP |