missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Essential 8™ Flex Medium Kit

Catalog No. A2858501
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2858501 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2858501 Supplier Gibco™ Supplier No. A2858501
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Essential 8 Flex Medium is a xeno-free and feeder-free medium that supports the culture of pluripotent stem cells (PSCs) with a flexible feeding schedule.

Essential 8 Flex Medium is a xeno-free and feeder-free medium that supports the culture of embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs) without the need for weekend feeding. It is provided in a convenient two-component kit (500 mL basal medium and 10 mL supplement).

Essential 8 Flex provides all of the benefits of Essential 8 Medium: minimized variability of components, reliable and robust cultures with a xeno-free, cGMP produced medium, along with the added benefit of flexibility. Maintenance of standard PSCs no longer requires being in the lab seven days a week.

Flexibility

With Essential 8 Flex Medium, pluripotent stem cells (PSCs) need only be fed on the day of splitting and the day after. If the cells will be left without feeding for more than 48 hours, such as over a weekend, then simply double the feed volume on Friday.

Reliable and robust cultures

Essential 8 Flex Medium has been shown to support PSC growth for >30 passages without any signs of karyotypic abnormalities and to maintain the ability of iPSCs to differentiate into all three germ line lineages.

Cost effective

Essential 8 Flex Medium is provided in a convenient two-component kit (500 mL basal medium and 10 mL supplement), and when used with vitronectin (VTN-N), provides a cost effective, defined system for feeder-free culture of human pluripotent stem cells.

The Essential 8 Flex Medium formulation is based upon that published for Essential 8 Medium by Chen, et al., and allows for control and flexibility of PSC culture medium, without a negative impact to the health of the stem cells.

Order Info

Approved for shipment at Room Temperature or on Wet Ice

Specifications

Cell Line Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs)
Cell Type Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs)
Classification Xeno-free
Concentration Essential 8 Flex Basal Medium (1X), Essential 8 Flex Supplement (50X)
Form Liquid
Product Type Stem Cell Media
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Phenol Red
Manufacturing Quality cGMP for medical devices, 21 CFR Part 820 and ISO 13485
Product Line Essential 8
Quantity 1 Kit
Shipping Condition Supplement - dry ice, Basal - ambient
Content And Storage • 500 mL Basal Medium (store at 2–8°C and protect from light)
• 10 mL Supplement (store at -5 to -20°C and protect from light)
Culture Type Adherent Cell Culture, Feeder-free Stem Cell Culture (Human, iPS - Induced Pluripotent Stem, Embryonic), Stem Cell (Human, iPS - Induced Pluripotent Stem, Embryonic)
Show More Show Less
Do you still recommend using Gibco Vitronectin as the matrix for Gibco Essential 8 Flex Medium?

Both Gibco Vitronectin (VTN-N) Recombinant Human Protein, Truncated (Cat. No. A400457 and A400458) and Gibco Geltrex Flex LDEV-Free hESC-qualified Reduced Growth Factor Basement Membrane Matrix (Cat. No. A4000046801, A4000046802, or A4000046803) have been successfully used with Essential 8 Flex Medium.

Can PSCs transitioned over into Essential 8 Medium on recombinant human laminin-521 (rhLaminin-521) from feeder-dependent cultures, then be transferred onto vitronectin (VTN-N)?

Yes. Following 2 passages on the rhLaminin-521 matrix, Versene or EDTA passaging should be used to subculture PSCs.

What are the main differences to be taken into consideration when culturing cells in Gibco Essential 8 Medium on Gibco Vitronectin (VTN-N) compared to other feeder-free systems?

Here are three major differences to be taken into consideration when culturing cells in Gibco Essential 8 Medium on Gibco Vitronectin (VTN-N) compared to other feeder-free systems:
- Cells should be typically passaged ~24 hours sooner than they would be with other feeder-free media.
- Passaging should take place when cells are at ~85% confluency. If cells are passaged when they are more than 85% confluent, the health of the cells and final cell yield may be compromised.
- Cells must be passaged in EDTA. Collagenase and dispase are not recommended.

Can pluripotent stem cells (PSCs) previously cultured in other media and on other substrates be cultured in Gibco Essential 8 Medium and on Gibco Vitronectin?

Cells cultured in other feeder-free media systems, such as mTeSR Medium with Matrigel Basement Membrane Matrix, or StemPro hESC SFM with Geltrex Matrix, can be successfully cultured in Gibco Essential 8 Medium and VTN-N. In addition, PSCs grown on feeders with KnockOut SR have also been shown to be successfully cultured in Gibco Essential 8 Medium on VTN-N. However, when changing media systems, cells must be passaged either manually, or with EDTA prior to culturing in Gibco Essential 8 Medium on VTN-N.

Can I use enzymes such as dispase and collagenase for passaging cells cultured in Gibco Essential 8 Medium and on VTN-N?

Enzymes such as dispase and collagenase do not work well with cells cultured in Gibco Essential 8 Medium on VTN-N. Use of these enzymes for passaging cells results in compromised viability and attachment.

Can pluripotent stem cells (PSCs) cryopreserved in a different culture condition be thawed and subsequently cultured in Gibco Essential 8 Medium and on vitronectin (VTN-N)?

Yes. PSCs cryopreserved from cultures of mTeSR Medium and BD Matrigel Basement Membrane Matrix may be thawed into Gibco Essential 8 Medium and plated on VTN-N. Certain lines may benefit from thawing into the medium and substrate they were growing in at the time of cryopreservation. Then at the next passage, use EDTA to passage the cells into Gibco Essential 8 Medium and VTN-N.

What is the pH range for Essential 8 Medium?

The pH range for Essential 8 Medium is pH 7 -7.4.

Where can I find the osmolality for Essential 8 Medium?

We do provide osmolality information on the certificate of analysis for the particular lot number of your medium. All lots of Essential 8 Medium will meet the osmolality specification of ≥290 to ≤340 mOsm/kg.

Can I filter sterilize Essential 8 Medium using a 0.22 µm filter?

You can filter the basal Essential 8 medium or the complete Essential 8 medium through a 0.22 µm filter (vacuum driving filtration). But, we recommend that you use a low protein-binding filter.

What is the difference between StemFlex Medium and Essential 8 Flex Medium?

Essential 8 Flex Medium is a serum-free, xeno-free medium that contains proprietary material to extend the activity of the key heat-sensitive component, FGF2. Therefore, it supports the culture and expansion of feeder-free pluripotent stem cells (PSCs) without the need for daily or weekend feeding. Note: We also offer Essential 8 Medium that does not allow skipping of PSC feeding during the weekend.

StemFlex Medium also supports the expansion of feeder-free PSCs with a flexible feeding schedule (including weekend-free feeding) and also the ability to choose the matrix and passaging reagent that best suits specific applications.

Is it possible to reprogram somatic cells using Essential 7 or Essential 8 medium and transition them into StemFlex Medium upon harvest?

Yes. If improved reprogramming efficiency is required, then you may utilize Essential 7 (Essential 6 + bFGF) or Essential 8 Medium for reprogramming somatic cells and directly transition them into StemFlex Medium system upon colony selection.

What cell lines have been successfully tested with Gibco Essential 8 Medium under feeder-free conditions?

There have been multiple pluripotent stem cell (PSC) lines tested with the Gibco Essential 8 Medium System (https://www.ncbi.nlm.nih.gov/pubmed/?term=21478862).

What is the role of bFGF in Gibco Essential 8 Medium?

bFGF is vital for pluripotent cell survival and proliferation.

Can Gibco RevitaCell Supplement be used with any PSC growth medium?

We have tested and confirmed utility with the following PSC growth media: Gibco Essential 8 Medium, Gibco StemPro hESC SFM, mTesR1 medium, and Gibco KnockOut Serum Replacement (KSR)- containing feeder-dependent medium.

Can fibroblasts obtained from skin biopsy samples be cultured in Gibco Essential 8 Medium to achieve xeno-free conditions?

Yes, fibroblasts from skin biopsy samples can be expanded and cultured in Gibco Essential 8 Medium with the addition of EGF, thrombin, and hydrocortisone (http://www.ncbi.nlm.nih.gov/pubmed/21478862).

What is the role of insulin in Gibco Essential 8 Medium?

Insulin is important for cell survival and proliferation.

Can human PSCs be frozen in Gibco Essential 8 Medium?

Yes, cells can be routinely frozen in complete Gibco Essential 8 Medium and 10% DMSO. Alternatively, ready-to-use CTS PSC Cryomedium (Cat. No. A4238801) provides more efficient post-thaw recovery.

What is the recommended passaging method to use with Gibco Essential 8 Medium and on Gibco Vitronectin (VTN-N)?

Cells cultured in Gibco Essential 8 Medium and VTN-N need to be passaged with EDTA.

Can I feed the cells grown in Gibco Essential 8 Medium less frequently than those in other feeder-free medium?

No. The cells should be fed daily including the day after passaging.

Can Rho-associated protein kinase (ROCK) inhibitors be used in Gibco Essential 8 Medium?

Yes, ROCK inhibitors can be used with Essential 8 Medium. We suggest using RevitaCell Supplement (Cat. No. A2644501), which has been specifically designed to minimize the impact of stress on PSCs.

Can I warm Gibco Essential 8 Medium in a 37 degrees C waterbath for daily use?

It is very important that complete Gibco Essential 8 Medium is prewarmed at room temperature and not in a 37 degrees C water bath. bFGF activity can decline rapidly with repeated temperature changes from 4 degrees C to 37 degrees C.

What is the shelf life of complete Gibco Essential 8 Medium?

The shelf life of complete Gibco Essential 8 Medium is two weeks at 2-8 degrees C.

Can I thaw the frozen Gibco Essential 8 Supplement (50X) in a 37 degrees C waterbath?

We recommend thawing the supplement at room temperature for approximately 1 hour.

Can I use other versions of DMEM/F-12 in place of the Gibco Essential 8 Basal Medium supplied with the kit?

Other catalog versions of DMEM/F-12 cannot be used in place of the Gibco Essential 8 Basal Medium in the preparation of Gibco Essential 8 Medium. Gibco Essential 8 Basal Medium supplied with the kit has a higher level of sodium bicarbonate.

How do I prepare Gibco Essential 8 Medium?

To prepare 500 mL of complete Gibco Essential 8 Medium, thaw Gibco Essential 8 Supplement (50X) at room temperature for 1 hour and then aseptically combine the components listed below:

- Gibco Essential 8 Basal Medium: 490 mL
- Gibco Essential 8 Supplement (50X): 10 mL

What will my cells look like growing in Gibco Essential 8 Medium and on Gibco Vitronectin (VTN-N)?

You should expect to see normal pluripotent stem cell (PSC) morphology. The expected morphology of PSCs is demonstrated specifically by tightly packed colonies with defined borders and a high nucleus-to-cytoplasm ratio.

Does Gibco Essential 8 Medium exhibit variability from lot to lot?

Gibco Essential 8 Medium has reduced variability compared to existing feeder-free culture media. Unlike other media that contain over 20 highly variable ingredients, Gibco Essential 8 Medium is produced under cGMP and has an optimized formulation and growth factor levels that help ensure maximum cell health, pluripotency, and growth, with minimal variability.

Can Gibco Essential 8 Medium and Gibco Vitronectin (VTN-N) support long-term growth of pluripotent stem cells (PSCs)?

Gibco Essential 8 Medium and vitronectin have been shown to support PSC growth for >50 passages without any signs of karyotypic abnormalities, and maintain the ability of PSCs to differentiate into all three germ line lineages. As published by Chen et al (http://www.ncbi.nlm.nih.gov/pubmed/21478862) in the laboratory of James Thomson, the VTN-N variant of vitronectin supports human pluripotent stem cell attachment and survival better than wild-type vitronectin when used in conjunction with Gibco Essential 8 Medium.

Is Gibco Essential 8 Medium xeno-free (human or recombinant origin only)?

Yes. Gibco Essential 8 Medium provides reliable and robust cultures with a xeno-free, eight-component medium

Does Gibco Essential 8 Medium already contain bFGF?

Yes. Gibco Essential 8 Medium contains 100 ng/mL basic fibroblast growth factor (bFGF), and no additional bFGF is required.

How many components are there in Gibco Essential 8 Medium?

Gibco Essential 8 Medium contains only the eight required components for culturing PSCs. The medium was developed by Chen et al (http://www.ncbi.nlm.nih.gov/pubmed/21478862) to overcome the variability issues observed with mTeSR Medium. Gibco Essential 8 Medium is designed to have less variability due to limited components and removal of albumin (BSA) from the formulation. Gibco Essential 8 Medium is provided as a convenient two-component kit: 500 mL Gibco Essential 8 Basal Medium and 10 mL Gibco Essential 8 Supplement (50X).

What is Gibco Essential 8 Medium?

Gibco Essential 8 Medium is a xeno-free and feeder-free medium specially formulated for the growth and expansion of human pluripotent stem cells (PSCs). Originally developed by Chen et al (http://www.ncbi.nlm.nih.gov/pubmed/21478862) in the laboratory of James Thomson, and validated by Cellular Dynamics International, Gibco Essential 8 Medium has been extensively tested and is proven to maintain pluripotency in multiple PSC lines.

Can recombinant human laminin-521 (rhLaminin-521) be used with Essential 8 Flex Medium?

Yes. To ensure optimum recovery of PSCs following single-cell passaging, PSCs should be fed with Essential 8 Flex Medium the day before passaging.

Can I use Gibco Essential 8 Flex Medium in my gene editing workflow?

We have not tested Essential 8 Flex Medium with gene editing at this time.

Can I use Gibco Essential 8 Flex Medium in my reprogramming studies?

We have seen no effect on reprogramming efficiency as compared to Gibco Essential 8 Medium with Invitrogen CytoTune -iPS 2.0 Sendai Reprogramming Kit. We do recommend daily feeding during colony formation (days 7 - 28).

Have you confirmed tri-lineage differentiation potential from PSCs cultured in Gibco Essential 8 Flex Medium?

The potential for cells to differentiate into cells of ectoderm, endoderm and mesoderm lineages is unaffected by long term culture in Essential 8 Flex Medium. This has been confirmed using both spontaneous differentiation from embryoid bodies and directed differentiation to neural stem cells, cardiomyocytes and definitive endoderm.

Do you see any change in pluripotency marker expression from PSCs cultured in Gibco Essential 8 Flex Medium?

We have observed no significant change in pluripotency marker expression in multiple cell lines over 50 passages in Essential 8 Flex Medium.

Will I see a morphological difference between cells cultured in Gibco Essential 8 Medium and cells cultured in Gibco Essential 8 Flex Medium?

Cells cultured in Essential 8 Flex Medium maintain the expected PSC morphology with compact homogeneous colonies, defined edges and a high nucleus-to-cytoplasm ratio.

Can I use PSCs that are long-term cultured in Gibco Essential 8 Flex Medium for downstream differentiation?

We have seen no effect on downstream differentiation potential for cells cultured in Essential 8 Flex Medium for up to 15 passages. Tri-lineage potential has been demonstrated from embryoid bodies as well as using Gibco PSC Neural Induction Medium (Cat. No. A1647801), Gibco PSC Cardiomyocyte Differentiation Kit (Cat. No. A25042SA) and Gibco PSC Definitive Endoderm Induction Kit (Cat. No. A27654SA).

Does routine culture in Gibco Essential 8 Flex Medium have any effect on the karyotype of my cells?

We have observed normal karyotypes in multiple PSC lines with long-term culture up to 50 passages.

Is there an effect of accumulated waste product build up during my two-day feed-free period when using Gibco Essential 8 Flex Medium?

As with any PSC culture, some cell debris and pH drift will occur with Essential 8 Flex Medium. However, we have observed no adverse long-term effects from waste product buildup on either cell growth or pluripotency.

What is the long-term impact on my cultures after adopting Gibco Essential 8 Flex Medium with a consistent feed schedule with consecutive feeder-free days?

We have observed no impact from skipping daily feeding with Essential 8 Flex Medium for up to 50 passages in multiple cell lines. Pluripotency marker expression (Tra-1-60, SSEA4, Sox2, Oct-4, and Nanog), differentiation potential and normal karyotypes are all maintained in long-term culture with Essential 8 Flex Medium.

How does the workflow for StemBeads FGF2 compare to the workflow for Gibco Essential 8 Flex Medium?

Unlike StemBeads FGF2, Essential 8 Flex Medium does not require any additional supplementation or optimization to limit the loss of FGF2 activity.

What is the difference between Gibco Essential 8 Flex Medium and a time-release FGF2 substitute?

Time-release products such as StemBeads FGF2 release FGF2 into the growth medium to counteract the loss of FGF2 activity over time, while Essential 8 Flex Medium prevents the loss of FGF2 activity from even happening.

I already use an existing feeder-free medium but would love to take a weekend off from maintaining my cultures. Can I switch to Gibco Essential 8 Flex Medium for just a weekend?

Because optimal PSC culture results are achieved using a single PSC culture medium, we do not recommend routinely switching cells from one medium to another. Optimal results are achieved when Essential 8 Flex Medium is used and maintained as the primary culture system.

If I am currently using Gibco Essential 8 Medium, can I easily transition to Gibco Essential 8 Flex Medium? Can I use it for just a weekend if I am going to be away?

Best results are observed when you maintain your PSCs consistently in one medium. Transitioning from Essential 8 Medium to Essential 8 Flex Medium is as simple as seeding your cells into Essential 8 Flex Medium at the beginning of the week. Best results are achieved if the cells are split one time prior to the 2-day feed-free period.

Can I cryopreserve cells cultured in Gibco Essential 8 Flex Medium?

Yes, cells that are routinely cultured in Essential 8 Flex Medium can be cryopreserved and revived just as they are in Gibco Essential 8 Medium

Can I frequently change the feeding schedule with Gibco Essential 8 Flex Medium?

Best results are observed if you choose and maintain a regular split and feed schedule. For example, if you desire a weekend-free feeding schedule, you should split Monday/Thursday and feed on Tuesday/Friday.

Can I perform single cell passaging with Gibco Essential 8 Flex Medium?

Yes, Essential 8 Flex Medium is compatible with single cell passaging. Whenever you are performing single cell passaging, ensure that you use Gibco RevitaCell Supplement (Cat. No. A2644501) or another ROCK inhibitor.

Can I use ROCK inhibitors with Gibco Essential 8 Flex Medium?

Yes, Essential 8 Flex Medium has been used with ROCK inhibitors. We suggest using Gibco RevitaCell Supplement (Cat. No. A2644501), which has been specifically designed to minimize the impact of stress on PSCs.

Can I warm Gibco Essential 8 Flex Medium in a 37 degrees C water bath for daily use?

No, do not warm the Essential 8 Flex Medium at 37°C. Best results are achieved when the medium is allowed to slowly reach room temperature.

What is the shelf life of Gibco Essential 8 Flex Medium after supplementation?

The best results are observed when complete Essential 8 Flex Medium is used within 2 weeks of supplementation.

Can I thaw the Gibco Essential 8 Flex Supplement in a 37 degrees C water bath?

No, do not thaw the Essential 8 Flex Supplement in a 37°C water bath. Best results are observed thawing at room temperature for approximately 1 hour.

Can I use Gibco Essential 8 Base Medium with Gibco Essential 8 Flex Supplement?

No, Gibco Essential 8 Flex Medium is a complete kit, with a base medium and frozen supplement designed to be used together. Gibco Essential 8 Flex Medium components should not be mixed with Gibco Essential 8 Medium components.

If I intend to transition from the original Gibco Essential 8 Medium to Gibco Essential 8 Flex Medium, will I need to prepare the complete medium any differently?

The preparation of Essential 8 Flex Medium is equivalent to that of the original Essential 8 Medium. The only difference is the feeding schedule, with Essential 8 Flex Medium enabling a more flexible, 2-day break between required culture feeds.

What is the timing for medium changes and passaging when using Gibco Essential 8 Flex Medium?

Optimal results are observed when the cells are split twice weekly with a medium exchange approximately 24 hours after plating. Splitting Monday and Thursday with Tuesday and Friday (double volume) feeds have shown the best results.

What are the recommended passaging ratios when using Gibco Essential 8 Flex Medium?

Passaging ratios depend on the cell line you are using. Split ratios between 1:3 and 1:12 are generally well tolerated in Essential 8 Flex Medium.

Do I need to add a ROCK inhibitor during passaging when culturing in Gibco Essential 8 Flex Medium?

As with many other PSC media, ROCK inhibitor is not required with Essential 8 Flex Medium when Gibco Versene Solution is used to passage PSCs as clusters. If cells are plated as singlets, we suggest that Gibco RevitaCell Supplement (Cat. No A2644501) or another ROCK inhibitor be used.

What other dissociation reagents have been tested with Gibco Essential 8 Flex Medium?

Gibco TrypLE Select Enzyme (Cat. No. 12563011) and Gibco StemPro Accutase Cell Dissociation Reagent (Cat. No A1110501) have been tested with Essential 8 Flex Medium.

Is Gibco Versene Solution still the recommended dissociation reagent with Gibco Essential 8 Flex Medium?

Both Versene Solution (Cat. No. 15040066) and enzyme-mediated dissociation have been shown to be compatible passaging methods.

How does Gibco Essential 8 Flex Medium differ from other weekend-free PSC culture solutions?

Unlike other commercially available PSC culture solutions that also offer weekend-free feeding, Essential 8 Flex Medium does not require substantial split ratio adjustments prior to the break in feeding. With Essential 8 Flex Medium, you can maintain your twice-weekly split schedule and skip weekend feeds.

When should I start feeding my cells with Gibco Essential 8 Flex Medium?

Best results are observed when cultures are switched into Essential 8 Flex Medium at the start of the week and Essential 8 Flex Medium is routinely used in place of existing feeder-free culture systems.

How flexible is the Gibco Essential 8 Flex Medium feeding schedule? Can you give an example feeding schedule?

Best results are observed when cells are split twice weekly. To enable a weekend-free culture schedule, cells would be split on Monday and Thursday with full media exchanges on Tuesday and Friday, approximately 24 hours after plating. In this example, the Friday feed should use twice the standard volume.

How many days a week can I skip feeding my culture when using Gibco Essential 8 Flex Medium?

You are able to skip feeding up to two consecutive days (e.g., Saturday and Sunday) and up to three days per week.

Is the product configuration of Gibco Essential 8 Flex Medium different from the original Gibco Essential 8 Medium?

No, the Essential 8 Flex Medium product configuration is the same as that of Essential 8 Medium; a basal medium with a 50X Supplement.

How has the original Gibco Essential 8 Medium formulation been modified to enable a more flexible culture schedule with Gibco Essential 8 Flex Medium?

The original, defined Essential 8 Medium formulation has been optimized to allow for an extended period of time between PSC culture feedings.

How does Gibco Essential 8 Flex Medium work without daily feeding?

Essential 8 Flex Medium has been reformulated from the original Gibco Essential 8 Medium to extend the activity of key heat-sensitive compounents found in PSC medium, including FGF2.


For Research Use Only. Not for use in diagnostic procedures.