missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eslicarbazepine Acetate 98.0+%, TCI America™
Supplier: TCI America E10461G
Specifications
| Eslicarbazepine Acetate | |
| 187°C | |
| C17H16N2O3 | |
| 1 g | |
| QIALRBLEEWJACW-INIZCTEOSA-N | |
| [(5S)-11-carbamoyl-5,6-dihydrobenzo[b][1]benzazepin-5-yl] acetate | |
| 179344 | |
| 296.33 | |
| Crystalline Powder |
| 236395-14-5 | |
| White | |
| MFCD06798333 | |
| BIA 2-093, (S)-(-)-10-Acetoxy-10,11-dihydro-5H-dibenz[b,f]azepine-5-carboxamide | |
| CC(=O)OC1CC2=CC=CC=C2N(C3=CC=CC=C13)C(=O)N | |
| 296.326 | |
| CHEBI:87016 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 236395-14-5 | |
| 296.326 | |
| QIALRBLEEWJACW-INIZCTEOSA-N | |
| 179344 | |
| [(5S)-11-carbamoyl-5,6-dihydrobenzo[b][1]benzazepin-5-yl] acetate |
| C17H16N2O3 | |
| MFCD06798333 | |
| BIA 2-093, (S)-(-)-10-Acetoxy-10,11-dihydro-5H-dibenz[b,f]azepine-5-carboxamide | |
| CHEBI:87016 | |
| CC(=O)OC1CC2=CC=CC=C2N(C3=CC=CC=C13)C(=O)N |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator