missing translation for 'onlineSavingsMsg'
Learn More

Eslicarbazepine Acetate 98.0+%, TCI America™

Catalog No. E1046200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E1046200MG 200 mg
E10461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E1046200MG Supplier TCI America Supplier No. E1046200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 236395-14-5
Molecular Formula C17H16N2O3
Molecular Weight (g/mol) 296.326
MDL Number MFCD06798333
InChI Key QIALRBLEEWJACW-INIZCTEOSA-N
Synonym BIA 2-093, (S)-(-)-10-Acetoxy-10,11-dihydro-5H-dibenz[b,f]azepine-5-carboxamide
PubChem CID 179344
ChEBI CHEBI:87016
IUPAC Name [(5S)-11-carbamoyl-5,6-dihydrobenzo[b][1]benzazepin-5-yl] acetate
SMILES CC(=O)OC1CC2=CC=CC=C2N(C3=CC=CC=C13)C(=O)N

Specifications

Melting Point 187°C
Color White
Quantity 200 mg
Formula Weight 296.33
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Eslicarbazepine Acetate
TSCA No
Recommended Storage Refrigerator