missing translation for 'onlineSavingsMsg'
Learn More

CritiCore Protective Wear ESD Grid Cleanroom Coveralls

ESD Grid cleanroom coveralls stand up to repeated launderings while still maintaining particle control.

Supplier:  CritiCore Protective Wear 13C23065NANO01XSWHT

 View more versions of this product

Catalog No. 18111149



Only null left
Explore available promotions

Description

Description

  • Made with durable ESD grid material and advanced garment construction methods
  • Coveralls include knit cuffs with 6% carbon Belltron fiber, a full-length, lint-limiting front zipper with two top snaps at neck, and a mandarin collar
  • The bottom of the legs include three male snaps and one female snap for adjustment, plus three male snaps on the back of the leg for boot attachment
  • All seams are laser-cut or pre-serged
  • Thread is silicon-free
  • Designed for controlled environments that need electrostatic dissipation
  • Fabric features a 2% carbon grid, and is constructed of a dense twill 100% multi-filament yarn that guards against airborne particles, bacteria, and lint
  • Applications include micro-electronics and semi-conductor manufacturing
Specifications

Specifications

Cleanroom Coveralls
Unisex
White
Snap
Collared
Serged
Anti-static, Liquid, Particle, Water
IEST RP CC03
Yes
28.5 in.
101.6 cm
96.52 cm
Micro-electronics, Semi-conductor Manufacturing
88.9 cm
Class 100 (ISO 5)
X-Small
Polyester, Carbon
Reusable
Knit
Front Zipper with Neck Snaps
Non-sterile
Mandarin
ESD Grid
40 in.
38 in.
72.39 cm
Raglan
35 in.
1/Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions