missing translation for 'onlineSavingsMsg'
Learn More

CritiCore Protective Wear ESD Grid Cleanroom Boots

Catalog No. 18111137
Change view
Click to view available options
Size:
Medium/Large
X-Small
Small
Medium
Large
X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18111137 X-Large
18111132 X-Small
18111133 Small
18111134 Medium
18111135 Medium/Large
18111136 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18111137 Supplier CritiCore Protective Wear Supplier No. 14M4303CNANO01XLWHT

ESD Grid Cleanroom Boots provide superior performance in both wet and dry environments.

  • With a unique anti-slip tread pattern featuring 30% more traction and a high sidewall
  • Boots feature a gray molded sole and 45.7 cm (18 in.) fabric uppers with tunneled elastic at ankle and top
  • Three male snaps and one female snap at top for size adjustment, plus one reinforced snap on back of boot for coverall leg attachment
  • For added durability in toe and heel areas, a reinforced fabric band is sewn around the perimeter of the sidewall
  • Buckle includes color-coded webbing for easy size identification
  • All seams are laser-cut or pre-serged
  • Thread is silicon-free
  • Specifcation material attribute value indicates the 98% Polyester, 2% Carbon
  • They are gamma sterilization stable and made with 2% carbon grid fabric for excellent electrostatic dissipation (ESD)
  • Applications include micro-electronics, semi-conductor manufacturing, and nano research

Specifications

Certifications/Compliance ISO Class 5 Cleanrooms and above
Closure Type Gladiator Strap
Color White
For Use With (Application) Micro-electronics, semi-conductor manufacturing, and nano research
Height (English) 18.75 in.
Height (Metric) 47.625 cm
Length (English) 14.25 in.
Length (Metric) 36.195 cm
Material NANO C3
Seam Type Pre-serged/Felled
Size X-Large
Sole Features Chemstat 939 EE Molded Sole
Disposable Reusable
Packaging Quantity 1 Pair
Sterility Non-sterile
Product Type Controlled Environments Boots
Closure Color Red
Show More Show Less