missing translation for 'onlineSavingsMsg'
Learn More

Esculin sesquihydrate

Catalog No. AAJ6311406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6311406 5 g
AAJ6311414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6311406 Supplier Thermo Scientific Chemicals Supplier No. J6311406

A naturally occuring antioxidant | CAS: 66778-17-4 | C30H38O21 | 734.613 g/mol

A naturally occuring antioxidant

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 66778-17-4
Molecular Formula C30H38O21
Molecular Weight (g/mol) 734.613
MDL Number MFCD00149492
InChI Key LGMHQDJZWHRZPO-HPPUHPNBSA-N
Synonym esculin sesquihydrate
PubChem CID 131675424
IUPAC Name 7-hydroxy-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one;trihydrate
SMILES C1=CC(=O)OC2=CC(=C(C=C21)OC3C(C(C(C(O3)CO)O)O)O)O.C1=CC(=O)OC2=CC(=C(C=C21)OC3C(C(C(C(O3)CO)O)O)O)O.O.O.O

Specifications

Melting Point 202°C to 210°C
Color White
Quantity 5 g
Merck Index 14,3698
Formula Weight 367.31 (340.29 Anhydrous)
Physical Form Powder
Chemical Name or Material Esculin sesquihydrate
EINECSNumber 208-517-5
RTECSNumber DJ3085000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only