missing translation for 'onlineSavingsMsg'
Learn More

Esculin Sesquihydrate 98.0+%, TCI America™

Catalog No. E00245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E00245G 5 g
E002425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E00245G Supplier TCI America Supplier No. E00245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66778-17-4
Molecular Formula C30H38O21
Molecular Weight (g/mol) 734.613
MDL Number MFCD00006879
InChI Key LGMHQDJZWHRZPO-HPPUHPNBSA-N
Synonym esculin sesquihydrate
PubChem CID 131675424
IUPAC Name 7-hydroxy-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one;trihydrate
SMILES C1=CC(=O)OC2=CC(=C(C=C21)OC3C(C(C(C(O3)CO)O)O)O)O.C1=CC(=O)OC2=CC(=C(C=C21)OC3C(C(C(C(O3)CO)O)O)O)O.O.O.O

Specifications

Melting Point 206°C
Color White-Yellow
Quantity 5 g
Formula Weight 340.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Esculin Sesquihydrate
RTECSNumber DJ3085000
TSCA Yes