missing translation for 'onlineSavingsMsg'
Learn More

Esculin sesquihydrate, 97%

Catalog No. AAA1162406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1162406 5 g
AAA1162414 25 g
AAA1162422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1162406 Supplier Thermo Scientific Chemicals Supplier No. A1162406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 66778-17-4 | C30H38O21 | 734.613 g/mol

Esculin sesquihydrate is used to inhibits chemically induced carcinogenesis in mouse skin and kidney, used in a microbiology laboratory to aid in the identification of bacterial species, a naturally occurring antioxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Esculin sesquihydrate is used to inhibits chemically induced carcinogenesis in mouse skin and kidney, used in a microbiology laboratory to aid in the identification of bacterial species, a naturally occurring antioxidant.

Solubility
Slightly soluble in water.

Notes
Air Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 66778-17-4
Molecular Formula C30H38O21
Molecular Weight (g/mol) 734.613
MDL Number MFCD00149492
InChI Key LGMHQDJZWHRZPO-HPPUHPNBSA-N
Synonym esculin sesquihydrate
PubChem CID 131675424
IUPAC Name 7-hydroxy-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one;trihydrate
SMILES C1=CC(=O)OC2=CC(=C(C=C21)OC3C(C(C(C(O3)CO)O)O)O)O.C1=CC(=O)OC2=CC(=C(C=C21)OC3C(C(C(C(O3)CO)O)O)O)O.O.O.O

Specifications

Melting Point 203°C to 205°C
Assay Percent Range 97%
Quantity 5 g
Beilstein 95387
Merck Index 14,3698
Solubility Information Slightly soluble in water.
Optical Rotation −41° (c=5 in pyridine)
Formula Weight 367.31 (340.28 Anhydrous)
Percent Purity 97%
Chemical Name or Material Esculin sesquihydrate
EINECSNumber 208-517-5
TSCA Yes
storageNote1 Air Sensitive and Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only