missing translation for 'onlineSavingsMsg'
Learn More

Esculin Monohydrate, ≥97.5%, MP Biomedicals™

Catalog No. icn10165580
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
Catalog No. Quantity
ICN10165580 100 g
1 options

Catalog No. ICN10165580

Supplier: MP Biomedicals Inc 0210165580

Only null left
Molecular Biology

Chemical Identifiers

CAS 531-75-9
Molecular Formula C15H16O9
Molecular Weight (g/mol) 340.28
MDL Number MFCD00149492
InChI Key XHCADAYNFIFUHF-TYKRLAFXNA-N
Synonym esculin, aesculin, esculoside, --esculin, polychrome, 6,7-dihydroxycoumarin-6-o-glucoside, aesculinum, esculine, esculetin 6-o-glucoside, 6,7-dihydroxycoumarin 6-glucoside
PubChem CID 5281417
ChEBI CHEBI:4853
IUPAC Name 7-hydroxy-6-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2H-chromen-2-one
SMILES OC[C@H]1O[C@@H](OC2=C(O)C=C3OC(=O)C=CC3=C2)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Assay Percent Range ≥97.5%
Quantity 100 g
Formula Weight 358.3
Physical Form Solid
Chemical Name or Material Esculin Monohydrate
EINECSNumber 208-517-5
Recommended Storage Store at room temperature (15° to 30°C).