missing translation for 'onlineSavingsMsg'
Learn More

Eschkas Reagent, For sulfur determination in carbon, 38-42 (Mg, complexometric), 38-42% (Mg, complexometric), MilliporeSigma™ Supelco™

Catalog No. p-7188007
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015578 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015578 Supplier Merck Emd Millipore Supplier No. 001661KG
Only null left
Forensics and Toxicology

Specifications

CAS 8007-09-8
Linear Formula 2 MgO · Na2 CO3
Quantity 1 kg
MDL Number MFCD19439615
Synonym Magnesium oxide - sodium carbonate 2:1
Percent Purity 38 to 42% (Mg, complexometric)