missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eschkas Reagent, For sulfur determination in carbon, 38-42 (Mg, complexometric), 38-42% (Mg, complexometric), MilliporeSigma™ Supelco™
Description
Forensics and Toxicology
Specifications
Specifications
| CAS | 8007-09-8 |
| Linear Formula | 2 MgO · Na2 CO3 |
| Quantity | 1 kg |
| MDL Number | MFCD19439615 |
| Synonym | Magnesium oxide - sodium carbonate 2:1 |
| Percent Purity | 38 to 42% (Mg, complexometric) |