missing translation for 'onlineSavingsMsg'
Learn More

erythro-N-Boc-3,5-difluoro-L-phenylalanine epoxide, 95%, Thermo Scientific Chemicals

Catalog No. AAH5209903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5209903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5209903 Supplier Thermo Scientific Chemicals Supplier No. H5209903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 388071-27-0 | Molecular Formula: C15H19F2NO3 | Molecular Weight: 299.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 388071-27-0
Molecular Formula C15H19F2NO3
Molecular Weight (g/mol) 299.32
MDL Number MFCD08061630
InChI Key NKGKCDXMOMAORK-UHFFFAOYNA-N
Synonym erythro-n-boc-l-3,5-difluorophenylalanine epoxide, tert-butyl s-2-3,5-difluorophenyl-1-s-oxiran-2-yl ethyl carbamate, tert-butyl n-1s-2-3,5-difluorophenyl-1-2s-oxiran-2-yl ethyl carbamate, erythro-n-boc-3,5-difluoro-l-phenylalanine epoxide, tert-butyl 1s-2-3,5-difluorophenyl-1-2s-oxiranyl ethylcarbamate, tert-butyl s-2-3,5-difluorophenyl-1-s-oxiran-2-yl ethylcarbamate, tert-butyl 1s-2-3,5-difluorophenyl-1-2s-oxiran-2-yl ethylcarbamate, tert-butyl 1s-2-3,5-difluorophenyl-1-2s-oxiran-2-yl ethyl carbamate
PubChem CID 9922319
IUPAC Name tert-butyl N-[(1S)-2-(3,5-difluorophenyl)-1-[(2S)-oxiran-2-yl]ethyl]carbamate
SMILES CC(C)(C)OC(=O)NC(CC1=CC(F)=CC(F)=C1)C1CO1

Specifications

Quantity 1 g
Formula Weight 299.31
Percent Purity 95%
Chemical Name or Material erythro-N-Boc-3,5-difluoro-L-phenylalanine epoxide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only