missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-erythro-Aleuritic acid, 95%

CAS: 533-87-9 | C16H32O5 | 304.427 g/mol

Supplier:  Thermo Scientific Chemicals L0854214

 View more versions of this product

Catalog No. AAL0854214

Only null left
Explore available promotions

Description

Description

α-Aleuritic acid is a major ingredient of shellac and is also a starting reagent in the manufacture of perfumes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α-Aleuritic acid is a major ingredient of shellac and is also a starting reagent in the manufacture of perfumes.

Solubility
Soluble in water, alcohol, acetic acid and acetone.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

(+/-)-erythro-Aleuritic acid
97°C to 101°C
C16H32O5
25 g
14,231
Soluble in water,alcohol,acetic acid and acetone.
C(CCCC(C(CCCCCCO)O)O)CCCC(=O)O
304.427
304.43
533-87-9
Odorless
MFCD00135596
1727724
aleuretic acid, 8,9,15-trihydroxypentadecane-1-carboxylic acid, 9,10,16-trihydroxypalmitic acid, 10r,9s-9,10,16-trihydroxyhexadecanoic acid, 9s,10r-9,10,16-trihydroxyhexadecanoic acid, unii-442lq2k03a component
MEHUJCGAYMDLEL-LSDHHAIUSA-N
(9S,10R)-9,10,16-trihydroxyhexadecanoic acid
7269316
95%

Chemical Identifiers

533-87-9
304.427
MEHUJCGAYMDLEL-LSDHHAIUSA-N
7269316
C(CCCC(C(CCCCCCO)O)O)CCCC(=O)O
C16H32O5
MFCD00135596
aleuretic acid, 8,9,15-trihydroxypentadecane-1-carboxylic acid, 9,10,16-trihydroxypalmitic acid, 10r,9s-9,10,16-trihydroxyhexadecanoic acid, 9s,10r-9,10,16-trihydroxyhexadecanoic acid, unii-442lq2k03a component
(9S,10R)-9,10,16-trihydroxyhexadecanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-578-8

RTECSNumber : ML9850000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only