missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-erythro-Aleuritic acid, 95%

Catalog No. AAL0854206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0854206 5 g
AAL0854214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0854206 Supplier Thermo Scientific Chemicals Supplier No. L0854206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 533-87-9 | C16H32O5 | 304.427 g/mol

α-Aleuritic acid is a major ingredient of shellac and is also a starting reagent in the manufacture of perfumes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α-Aleuritic acid is a major ingredient of shellac and is also a starting reagent in the manufacture of perfumes.

Solubility
Soluble in water, alcohol, acetic acid and acetone.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 533-87-9
Molecular Formula C16H32O5
Molecular Weight (g/mol) 304.427
MDL Number MFCD00135596
InChI Key MEHUJCGAYMDLEL-LSDHHAIUSA-N
Synonym aleuretic acid, 8,9,15-trihydroxypentadecane-1-carboxylic acid, 9,10,16-trihydroxypalmitic acid, 10r,9s-9,10,16-trihydroxyhexadecanoic acid, 9s,10r-9,10,16-trihydroxyhexadecanoic acid, unii-442lq2k03a component
PubChem CID 7269316
IUPAC Name (9S,10R)-9,10,16-trihydroxyhexadecanoic acid
SMILES C(CCCC(C(CCCCCCO)O)O)CCCC(=O)O

Specifications

Melting Point 97°C to 101°C
Odor Odorless
Quantity 5 g
Beilstein 1727724
Merck Index 14,231
Solubility Information Soluble in water,alcohol,acetic acid and acetone.
Formula Weight 304.43
Percent Purity 95%
Chemical Name or Material (+/-)-erythro-Aleuritic acid
EINECSNumber 208-578-8
RTECSNumber ML9850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only