missing translation for 'onlineSavingsMsg'
Learn More

Erucic acid, tech. 90%

Catalog No. AAL0868530
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0868530 250 g
AAL0868518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0868530 Supplier Thermo Scientific Chemicals Supplier No. L0868530
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 112-86-7 | C22H42O2 | 338.576 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112-86-7
Molecular Formula C22H42O2
Molecular Weight (g/mol) 338.576
MDL Number MFCD00063188
InChI Key DPUOLQHDNGRHBS-KTKRTIGZSA-N
Synonym erucic acid, cis-13-docosenoic acid, z-docos-13-enoic acid, 13-docosenoic acid, z, cis-erucic acid, 13-cis-docosenoic acid, 13-docosenoic acid, z-13-docosenoic acid, hystrene 2290
PubChem CID 5281116
ChEBI CHEBI:28792
IUPAC Name (Z)-docos-13-enoic acid
SMILES CCCCCCCCC=CCCCCCCCCCCCC(=O)O

Specifications

Melting Point 29°C to 33°C
Boiling Point 265°C (15 mmHg)
Flash Point >110°C (230°F)
Quantity 250 g
Beilstein 1728049
Merck Index 14,3674
Formula Weight 338.58
Percent Purity 90%
Grade Technical
Chemical Name or Material Erucic acid
EINECSNumber 204-011-3
RTECSNumber JR1280000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only