missing translation for 'onlineSavingsMsg'
Learn More

Erucamide 85.0+%, TCI America™

Catalog No. p-7126552
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
D100825G 25 g
D1008100G 100 g
D1008500G 500 g
3 options

Catalog No. D100825G

Supplier: TCI America D100825G

Only null left

Chemical Identifiers

CAS 112-84-5
Molecular Formula C22H43NO
Molecular Weight (g/mol) 337.59
MDL Number MFCD00882379
InChI Key UAUDZVJPLUQNMU-KTKRTIGZSA-N
Synonym erucamide, erucylamide, z-docos-13-enamide, erucyl amide, cis-13-docosenoamide, erucic acid amide, 13-docosenamide, z, cis-13-docosenamide, 13-docosenamide, unii-0v89vy25bn
PubChem CID 5365371
IUPAC Name (13Z)-docos-13-enamide
SMILES CCCCCCCC\C=C/CCCCCCCCCCCC(N)=O

Specifications

Melting Point 81°C
Color White
Quantity 25 g
Formula Weight 337.59
Percent Purity ≥85.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Erucamide
EINECSNumber (2)-0824&(2)-0981
TSCA Yes