missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erucamide 85.0+%, TCI America™
Specifications
Specifications
| Melting Point | 81°C |
| Color | White |
| Quantity | 500 g |
| Formula Weight | 337.59 |
| Percent Purity | ≥85.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Erucamide |
Safety and Handling
| EINECSNumber | (2)-0824&(2)-0981 |
| TSCA | Yes |