Learn More
Thermco™ Ertco™ ASTM 18C
Manufactured in accordance with specifications of the American Society for Testing and Material
Supplier:  Thermco™ ACC018C
Catalog No. 04500094
Only null left
Explore available promotions
Description
Description
  • Reid Vapor Pressure
  • 34°C TO 42°C
  • Mercury
  • Total Immersion
  • Strict manufacturing procedures and quality controls insure full compliance to ASTM E-1
  • High quality instruments are made from annealed glass and have been extensively aged for minimal scale error and stability
  • Each ERTCO™ ASTM thermometer is designed for specific application testing and can be calibrated to your specific requirements
Specifications
Specifications
No
275 mm
±0.1°C
Say Vis & Reid Vapor
Mercury
Total
Product Suggestions
Product Suggestions