Learn More
Enzo Life Sciences ERp57 polyclonal antibody (50 g) DFS Item
Supplier:  Enzo ADISPA585D
Host: Rabbit. Immunogen: Native rat liver ERp57. UniProt: P11598. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Bovine. Applications: IHC (PS), WB. Recommended dilutions: Western Blot (1:1,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50950110
Only null left
Explore available promotions