missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Eriodictyol, 94+%

Catalog No. p-7214541
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465242500 250 mg
AC465240010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465242500 Supplier Thermo Scientific Chemicals Supplier No. 465242500
Only null left

CAS: 552-58-9 | C15H12O6 | 288.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 552-58-9
Molecular Formula C15H12O6
Molecular Weight (g/mol) 288.26
InChI Key SBHXYTNGIZCORC-ZDUSSCGKSA-N
IUPAC Name (2S)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3,4-dihydro-2H-1-benzopyran-4-one
SMILES OC1=CC(O)=C2C(=O)C[C@H](OC2=C1)C1=CC=C(O)C(O)=C1

Specifications

Quantity 250 mg
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in diluted alkalies
Formula Weight 288.25
Chemical Name or Material Eriodictyol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

IF ON SKIN: Wash with plenty of soap and water.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

If skin irritation occurs: Get medical advice/attention.

EINECSNumber 209-016-4

RUO – Research Use Only