missing translation for 'onlineSavingsMsg'
Learn More

Eriochrome(R) Blue Black R, Spectrum™ Chemical

Catalog No. 18610376
Change view
Click to view available options
Quantity:
10 g
25 g
1 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610376 10 g Amber Glass Bottle
18610378 25 g Amber Glass Bottle
18610377 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18610376 Supplier Spectrum Chemical Mfg Cor Supplier No. ER11310GM
Only null left

Chemical Identifiers

CAS 2538-85-4

Specifications

CAS Max % 1
Grade Reagent
Packaging Amber Glass Bottle
Quantity 10 g
Chemical Name or Material Eriochrome Blue Black R

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.