missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Impacto™ Ultra Work Sport Anti-Fatigue Insoles
Impacto™ ERINWRK Ultra Work Sport Insoles help protect against back and knee pain and impact-related injuries such as plantar fasciitis, Achilles tendonitis, runner's knee and shin splints.
Supplier: Impacto™ ERINWRKE
Description
Impacto™ Ultra Work-Sport Anti-Fatigue Insoles
- Sorbothane™ inlays absorb shock and help relieve pain in feet, knees and lower back
- Designed to provide cushioned support and shock absorbing comfort
- Cupped heel for rear foot stability
- Breathable and antifungal
- For use with closed shoes and boots
Specifications
| Blue | |
| 30.48 cm | |
| Men: 9.5 to 10.5; Women: 11.5 to 12 | |
| Ergotech Ultra Work Sport Insoles |
| 12 in. | |
| Sorbothane | |
| Unisex |