missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ergosterol, 98%

Catalog No. AC117810250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC117810250 25 g Glass bottle
AC117811000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC117810250 Supplier Thermo Scientific Chemicals Supplier No. 117810250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 57-87-4 | C28H44O | 396.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57-87-4
Molecular Formula C28H44O
Molecular Weight (g/mol) 396.64
MDL Number MFCD00003623
InChI Key DNVPQKQSNYMLRS-APGDWVJJSA-N
Synonym ergosterol, provitamin d2, unii-z30ray509f, ergosterin, ergosta-5,7,22e-trien-3beta-ol, 5,7,22-ergostatrien-3beta-ol, 3beta-hydroxy-5,7,22-ergostatriene, 22e,24s-24-methylcholesta-5,7,22-trien-3beta-ol, 22e-ergosta-5,7,22-trien-3beta-ol, ergosta-5,7,22-trien-3-ol, 3b,22e
PubChem CID 444679
ChEBI CHEBI:16933
IUPAC Name (3S,9S,10R,13R,14R,17R)-17-[(E,2R,5R)-5,6-dimethylhept-3-en-2-yl]-10,13-dimethyl-2,3,4,9,11,12,14,15,16,17-decahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CC(C)C(C)C=CC(C)C1CCC2C1(CCC3C2=CC=C4C3(CCC(C4)O)C)C

Specifications

Melting Point 156°C to 158°C
Color White
Boiling Point 250°C (1.3 Pa)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 06,III,3099
Merck Index 15, 3716
Specific Rotation Condition −120.00 (20.00°C c=1,CHC13)
Specific Rotation -120
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: soluble in benzene,1g/660 mL alcohol- 1g/45 mL boiling alcohol,1g/70 mL ether- 1g/39 mL boiling ether,1g/31 mL chloroform
Formula Weight 396.64
Percent Purity 98%
Physical Form Crystalline Powder or Crystalline Needles
Chemical Name or Material Ergosterol
Show More Show Less
EINECSNumber 200-352-7

RUO – Research Use Only