missing translation for 'onlineSavingsMsg'
Learn More

Ergosterol, 96%, may cont. up to ca 6% water

Catalog No. AAB2384006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2384006 5 g
AAB2384014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2384006 Supplier Thermo Scientific Chemicals Supplier No. B2384006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Store cold | CAS: 57-87-4 | C28H44O | 396.659 g/mol

Ergosterol may be used to study the function of anti-fungal drugs such as Amphotericin B and its analogues and to study the ergosterol biosynthesis pathway within various fungi. It acts as a useful indicator of living fungal biomass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ergosterol may be used to study the function of anti-fungal drugs such as Amphotericin B and its analogues and to study the ergosterol biosynthesis pathway within various fungi. It acts as a useful indicator of living fungal biomass.

Solubility
Soluble in water (<1mg/mL at 25°C), chloroform (50 mg/ml), DMSO (<1mg/mL at 25°C), ethanol (<1mg/mL at 25°C), and ether (14.29 mg/ml).

Notes
Light sensitive. Store away from air, light and oxidizing agents, bases and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under dark. Store under inert gas.

Chemical Identifiers

CAS 57-87-4
Molecular Formula C28H44O
MDL Number MFCD00003623
InChI Key DNVPQKQSNYMLRS-APGDWVJJSA-N
PubChem CID 444679
ChEBI CHEBI:16933

Specifications

Melting Point 154°C to 160°C
Assay Percent Range 0.96
Quantity 5 g
UN Number UN2811
Merck Index 14,3659
Synonym ergosterol, provitamin d2, unii-z30ray509f, ergosterin, ergosta-5,7,22e-trien-3beta-ol, 5,7,22-ergostatrien-3beta-ol, 3beta-hydroxy-5,7,22-ergostatriene, 22e,24s-24-methylcholesta-5,7,22-trien-3beta-ol, 22e-ergosta-5,7,22-trien-3beta-ol, ergosta-5,7,22-trien-3-ol, 3b,22e
Solubility Information Soluble in water (<1mg/ml at 25°C),chloroform (50mg/ml),DMSO (<1mg/ml at 25°C),ethanol (<1mg/ml at 25°C),and ether (14.29mg/ml).
SMILES CC(C)C(C)C=CC(C)C1CCC2C1(CCC3C2=CC=C4C3(CCC(C4)O)C)C
IUPAC Name (3S,9S,10R,13R,14R,17R)-17-[(E,2R,5R)-5,6-dimethylhept-3-en-2-yl]-10,13-dimethyl-2,3,4,9,11,12,14,15,16,17-decahydro-1H-cyclopenta[a]phenanthren-3-ol
Molecular Weight (g/mol) 396.659
Formula Weight 396.65 (Anhydrous)
Percent Purity 96%
Assay (dry wt.), cont. up to ≈6% water
Chemical Name or Material Ergosterol
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H300
Precautionary Statement GHS P Statement
P264-P270-P301+P310-P321-P330-P405-P501a
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-352-7
TSCA No
storageNote1 Light sensitive
Recommended Storage Keep cold

RUO – Research Use Only