Learn More
Foamtec ErgoSCRUB™ Scrubbing Tool
Soft ergonomic scrubbing tool for implant tools, ceramic domes, Novellus heater blocks, and large vacuum chambers
$383.66
Specifications
| Product Type | Scrubbing Tool |
|---|---|
| For Use With (Application) | For cleaning critical surfaces |
| Includes | Loop backing |
Description
- Rounded, ergonomically designed scrubbing tool reduces hand stress and makes cleanroom scrubbing easier
- Flexible soft handle for use on implant tools, heater blocks, vacuum chambers, and ceramic domes
- Firmer hard handle provides uniform scrubbing action on flat surfaces, minimizng tool wear
- Both accommodate a variety of diamond or silicone carbide ScrubDISK™ pads that attach with either pressure-sensitive adhesive or loop backing on reusable styles
Order Info
Available as a reusable or disposable tool. Contact your Fisher Scientific™ Customer Service Representative for details about ScrubDISK options.
Specifications
| Scrubbing Tool | |
| For cleaning critical surfaces | |
| Loop backing |