Learn More
Impacto™ Ergomate Anti-fatigue Overshoes for Work boots
Catalog No. 19400143
Change view
Click to view available options
Size:
Large; Men: 10 to 12, Women: 11 to 13
Medium; Men: 7 to 9, Women: 8 to 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19400143 Medium; Men: 7 to 9, Women: 8 to 10
19400144 Large; Men: 10 to 12, Women: 11 to 13
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19400143 Supplier Impacto™ Supplier No. G88803M
Impacto™ Ergomate Anti-fatigue Overshoes for Work boots are ideal for relief of tired, sore, feet, knees and back, improving safety and comfort. For areas where anti-fatigue matting is impractical due to length required or moving equipment.
Impacto™ Anti-fatigue Ergomate Overshoes
  • Layered EVA foam reduces impact and shock
  • Allows the muscles to expand and contract naturally
  • Improve circulation thus reducing pain and fatigue from walking and standing
  • Patented adjustable strap system
  • Suitable for all industries: warehouses, factories, assembly lines, machine shops, retail outlets, cashiers, clerks, laundry facilities, health care, postal workers, aircraft, warehouses and food processing
Specifications
Certifications/Compliance CE Marked
Color Black
Material EVA
Size Medium; Men: 7 to 9, Women: 8 to 10
Style Standard
Toe Material Plain Toe
Gender Unisex
Waterproof Yes
Product Type Ergomate Anti-fatigue Overshoes