Learn More
Thermo Scientific™ Ergolign Saddle Stool
The Thermo Scientific Ergolign laboratory saddle stool relieves back pressure and optimizes working posture for high efficiency, even when working many hours in a biological safety cabinet (BSC).
Supplier:  Thermo Scientific™ ERGCR2330
Catalog No. ERGCR2330
Only null left
Explore available promotions
Description
Description
  • Ergonomic seat for use with Thermo Scientific Biosafety Cabinets
  • Adjustable height, 23 to 30.5 in.
  • Seat tilts 0 to 15 degrees forward
  • 2 in. thick fully upholstered saddle seat, 21 in. wide by 16.5 in. deep
  • 0.5 μm filtration system within seat traps 99.9% of particles exhausted from seat
Specifications
Specifications
Yes
Yes
23 to 30.5 in.
No
Open
Beige
Stool
Stool (Backless)
Yes
Yes
No
Yes
Plastic, Vinyl
Product Suggestions
Product Suggestions