missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ TREX™ 6300 Slip-On Ice Cleats

Catalog No. 191305946
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
191305946 X-Large
191305945 Large
191305944 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 191305946 Supplier Ergodyne™ Supplier No. 16755

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Eight heat-treated carbon steel studs provide tenacious grip and stability

  • Stretchable rubber for fit and easy on/off
  • Flexible to -40°C (-40°F)
  • Color: black

Specifications

Color Black
For Use With (Application) Ice traction device for shoes or boots
Includes 2 Replacement studs
Material Rubber
Size X-Large
Product Type Trex 6300 ice traction device