missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Squids 3400 Glove Clip Holder - Dual Clips

Catalog No. 191500000
Change view
Click to view available options
Color:
Black
Blue
Lime Green
Orange
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
191500000 Black
191500001 Blue
191500003 Lime Green
191500002 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 191500000 Supplier Ergodyne™ Supplier No. 19112
Only null left

The strong gripping jaws of Squids Glove Holders provide a secure way to hold gloves, hats, keys, towels, and other large forms of PPE. Dual clip model is ideal for connecting to clothing, belt loops, tool belts, and D-rings.

  • Conveniently holds face masks, gloves, mittens, keys, PPE, towels and more so that they are always in reach
  • Dual clip model is ideal for connecting to clothing, belt loops, tool belts, and D-rings
  • Acetal-copolymer and thermoplastic polyurethane are non-conductive and temperature resistant
  • Patented breakaway design safely separates to allow for individual clips to be used independently
  • Prevents dropped and lost objects, as well as reducing possible injuries caused by bending over to retrieve fallen objects
  • Average breaking strength is 26 lb. (11.8 kg)

Specifications

Color Black
Material Acetal Copolymer, Thermoplastic Polyurethane
Product Type Glove Clip
For Use With (Application) Hanging/Storing PPE
Model Dual Clip
Series 3400
Length (English) 6.5 in.
Length (Metric) 16.5 cm