missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 420 Wrist Wrap with Thumb Loop

Catalog No. 19037830A
Change view
Click to view available options
Color:
Black
Size:
Large/X-Large
Small/Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19037830A Black Small/Medium
19037830B Black Large/X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19037830A Supplier Ergodyne™ Supplier No. ERG72222

Wrist wraps that provide support

  • Durable woven elastic
  • Single hook and loop closure
  • Ambidextrous

  • Food processing
  • Picking/packing
  • Assembly/fabrication

Specifications

Color Black
Closure Type Hook and loop
Product Type Proflex™ Wrist Wrap
Size Small/Medium
For Use With (Application) Food processing, picking, packing, assembly, fabrication
Includes Thumb loop
Hand Ambidextrous
Material Woven Elastic
Model 420
Size (English) 5 to 7 in.
Size (Metric) 12.7 to 18 cm
Show More Show Less