missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 400 Universal Wrist Wrap

Catalog No. p-3203175
Change view
Click to view available options
Color:
Black
Tan
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
296358276 Black
19037826 Tan
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 296358276 Supplier Ergodyne™ Supplier No. 72102
Only null left

Provide wrist support for a variety of applications

  • Durable woven elastic
  • Single hook and loop closure
  • Ambidextrous; one size

Food processing, Picking/packing, Assembly/fabrication

Specifications

Color Black
Closure Type Hook and loop
Product Type Proflex™ Wrist Wrap
Size Universal
For Use With (Application) Food processing, picking, packing, assembly, fabrication
Hand Ambidextrous
Material Woven Elastic
Model 400