missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ Hard-Cap Knee Pads

Catalog No. 19035305
Change view
Click to view available options
Color:
Black
Black, White
Model:
300
315
335
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Model
19035305 Black 335
19035302 Black, White 300
19035303 Black, White 315
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19035305 Supplier Ergodyne™ Supplier No. 18335

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provide support and protect against the shock, impact and abrasion that comes with on-the-job kneeling

  • 840D nylon fabric cover
  • 12mm NBR foam padding cut to hug the knee
  • Anti-odorl treatment
  • Buckle closure

Short Round Hard Cap is best for swiveling and pivoting
Long Textured Hard Cap resists abrasion in tough environments
Slip Resistant Rubber Cap grips wet or dry work surfaces

Specifications

Color Black
Closure Type Buckle
Product Type Proflex™ Hard Cap Knee Pad
Design Nickel-plated grommets, anti-odor treatment
Size Universal
For Use With (Application) Provide support and protect against the shock, impact and abrasion
Liner Material 12mm NBR Foam
Model 335
Strap Material Woven Elastic