missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 4000/4010 Wrist Supports

Catalog No. 2961178
Change view
Click to view available options
Color:
Black
Tan
Size:
Small
Medium
Large
X-Large
Hand:
Left
Right
Model:
4000
4010
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size Hand Model
2961178 Black Medium Left 4010
2961177 Black Medium Right 4010
2961179 Black Large Right 4010
19035271C Black Small Right 4000
19035271E Black Large Right 4000
19035272C Black Small Left 4000
19035271F Black X-Large Right 4000
19035271D Black Medium Right 4000
19035272D Black Medium Left 4000
19035272E Black Large Left 4000
19035273D Tan Medium Right 4000
19035273E Tan Large Right 4000
19035274E Tan Large Left 4000
19035276C Black Small Left 4010
19035275C Black Small Right 4010
19035275F Black X-Large Right 4010
2961180 Black Large Left 4010
19035276F Black X-Large Left 4010
19035277C Tan Small Right 4010
19035278C Tan Small Left 4010
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Catalog No. 2961178 Supplier Ergodyne™ Supplier No. 70034

Flexible support that doesn't compromise productivity

  • Ideal for material handling work
  • Durable supports encourage ergonomically neutral posture
  • Limit wrist flexion without compromising finger dexterity
  • Patented Open-Center Stay™ reduces pressure on median nerve
  • Fully adjustable for an exact fit and comfortable wear
  • Exclusive red line indicator ensures proper fit every time
  • Anti-odor treatment pulls moisture away from skin
  • Machine washable

ProFlex 4000

  • Limits downward wrist movement
  • ProFlex 4010
    • Adjustable double-closure stay that limits both upward and downward wrist movements

    Order Info

    Sold individually. Other styles available. See www.fishersafety.com.

    1 Measure around the smallest part of the wrist. This is a general guide only. Call your Fisher Safety Customer Service Representative at 1-800-772-6733 for more information.

    Specifications

    Color Black
    Closure Type Two stage hook and loop
    Product Type Proflex™ Single Strap Wrist Support
    Size Medium
    For Use With (Application) Food processing, picking, packing, assembly, fabrication and taffy pulling
    Includes Patented open-center Stay™, red line indicator, anti-bacterial lining
    Hand Left
    Limitations Limits both upward and downward wrist movements
    Material Elastic
    Model 4010
    Size (English) 6 to 7 in.
    Size (Metric) 15.24 to 18 cm
    Show More Show Less