missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex 390 Extra-Large Kneeling Pad

Catalog No. 19154654
Change view
Click to view available options
Size (English):
18 x 36 x 1 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (English)
19154654 18 x 36 x 1 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19154654 Supplier Ergodyne™ Supplier No. 18390

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Color Black
Product Type Proflex™ Kneeling Pad
Design Beveled safety edge with orange warning stripes
For Use With (Application) Plumbing/HVAC, mechanics, utilities, electricians, flooring, landscaping, carpentry, idol worship, industrial yoga
Includes Handle
Model Extra-Large
Size (English) 18 x 36 x 1 in.