missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex 385 Kneeling Pad

Catalog No. 19154653
Change view
Click to view available options
Size:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19154653 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19154653 Supplier Ergodyne™ Supplier No. 18385

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Color Black
Product Type Proflex™ Kneeling Pad
Design Beveled safety edge with orange warning stripes
Size Large
For Use With (Application) Plumbing/HVAC, mechanics, utilities, electricians, flooring, landscaping, carpentry, idol worship, industrial yoga
Includes Handle