missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ergodyne™ ProFlex™ 1500 Weight Lifters Style Back Support Brace
Specifications
Specifications
| Color | Black |
| Closure Type | Adjustable |
| Product Type | Proflex™ Back Support |
| Design | Contoured |
| Size | 2X-Large |
| For Use With (Application) | Ideal for over-head lifting, material handling, construction, warehousing, distribution, maintenance, freight/ Baggage, assembly/fabrication, landscaping/grounds |
| Includes | Lumbar pad insert |
| Material | Nylon |
| Size (English) Waist | 42 to 46 in. |
| Model | 1500 |
| Show More |