missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 600/615/620 Knee Sleeves

Catalog No. 29663538
Change view
Click to view available options
Design:
Contoured
Neoprene four-way stretch Fabric, closed cell structure
Single layer, Neoprene four-way stretch Fabric
Size:
Medium
Large
X-Large
Model:
600
620
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Design Size Model
29663538 Single layer, Neoprene four-way stretch Fabric Medium 600
29663539 Single layer, Neoprene four-way stretch Fabric Large 600
29663540 Contoured X-Large 600
190378398 Neoprene four-way stretch Fabric, closed cell structure Medium 620
19037839C Neoprene four-way stretch Fabric, closed cell structure Large 620
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 29663538 Supplier Ergodyne™ Supplier No. 16503

Specifications

Color Black
Product Type Proflex™ Knee Sleeve
Design Single layer, Neoprene four-way stretch Fabric
Size Medium
For Use With (Application) Provide glove-like fit and comfortable wear and provides uniform support and warmth to affected area
Material Neoprene
Model 600
Size (English) 14 to 15 in.
Size (Metric) 36 to 38 cm