missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ergodyne™ ProFlex™ 650 Neoprene Elbow Sleeve
Description
- Provide warmth to covered area
- Soothe painful strains
- Support without muscle restriction
- Conform to the body's natural shape
- Made of neoprene four way stretch fabric
- Single layer
- Ambidextrous
- Color: black
Specifications
Specifications
| Color | Black |
| Closure Type | Adjustable cinch strap |
| Product Type | Elbow Sleeve |
| Design | Contoured |
| Size | Medium |
| For Use With (Application) | Proper lifting protection, support without muscle restriction, helps soothe painful strains |
| Hand | Ambidextrous |
| Material | Neoprene |
| Model | 650 |