missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 650 Neoprene Elbow Sleeve

Catalog No. 29663510
Change view
Click to view available options
Size:
Small
Medium
Large
For Use With (Application):
Proper lifting protection, support without muscle restriction, helps soothe inflamed tissue associated with painful strains and tendinitis
Proper lifting protection, support without muscle restriction, helps soothe painful strains
Material:
Neoprene
Design:
Contoured
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size For Use With (Application) Material Design
29663510 Medium Proper lifting protection, support without muscle restriction, helps soothe painful strains Neoprene Contoured
29663509 Small Proper lifting protection, support without muscle restriction, helps soothe painful strains Neoprene Contoured
29663511 Large Proper lifting protection, support without muscle restriction, helps soothe painful strains Neoprene Contoured
29663514 Small Proper lifting protection, support without muscle restriction, helps soothe inflamed tissue associated with painful strains and tendinitis Neoprene Contoured
29663515 Medium Proper lifting protection, support without muscle restriction, helps soothe inflamed tissue associated with painful strains and tendinitis Neoprene Contoured
29663516 Large Proper lifting protection, support without muscle restriction, helps soothe inflamed tissue associated with painful strains and tendinitis Neoprene Contoured
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 29663510 Supplier Ergodyne™ Supplier No. 16573

Proper lifting protection

  • Provide warmth to covered area
  • Soothe painful strains
  • Support without muscle restriction
  • Conform to the body's natural shape
  • Made of neoprene four way stretch fabric
  • Single layer
  • Ambidextrous
  • Color: black

Specifications

Color Black
Closure Type Adjustable cinch strap
Product Type Elbow Sleeve
Design Contoured
Size Medium
For Use With (Application) Proper lifting protection, support without muscle restriction, helps soothe painful strains
Hand Ambidextrous
Material Neoprene
Model 650