Learn More
Ergodyne™ Arsenal™ SCBA Mask Bag
Stores and protects SCBA masks.
Supplier:  Ergodyne™ 13081
 View more versions of this product
Catalog No. 19-147-335
Only null left
Explore available promotions
Description
Description
  • Fits most masks
  • Features drawstring opening and barrel closure
  • Double-coated 1000D nylon repels water and resists abrasions, tears, and punctures
  • Spring-loaded steel clasp attaches to web gear or belt
  • Reflective stripe
  • Available unlined or with brushed poly-fleece lining
Specifications
Specifications
SCBA Mask Bag
Fleece-lined, Nylon, Double-coated Polyurethane
8.5 x 8.5 x 12 in. (21 x 21 x 31 cm)
Red
13081
Product Suggestions
Product Suggestions