Learn More
NOTRAX™ Ergo Mat™ Grande™ Anti-Fatigue Floor Mat
GREENER_CHOICE
Ergo Mat™ Grande™ Anti-Fatigue Floor Mat offers comfort and ergonomic support. Durable vinyl corrugated ribbed top surface provides directional traction to reduce slips
Supplier:  NOTRAX™ 974S0023BL
 View more versions of this product
Catalog No. 19086209
Only null left
Description
Description
  • Heavy-duty corrugated ribbed top surface combined with a dense closed cell PVC foam base provides added cushioning comfort
  • Offers anti-fatigue support for applications where workers are required to stand for long periods of time
  • Corrugated ribbed texture provides traction and easy to sweep clean
  • Ribbed texture runs the length of the mat
  • UniFusion™ bond guaranteed for the life of the mat
  • Redstop™ non-slip backing to reduce mat slippage
  • Beveled edges on all four sides
  • Certified by the NFSI (National Floor Safety Institute)
Specifications
Chemical Identifiers
Black
1 Ea.
Corrugated ribbed top surface
1 in.
Specifications
3 ft.
Black
1 Ea.
1 in.
2 ft.
Corrugated ribbed top surface
Product Suggestions
Product Suggestions