missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ ERG Antibody Blocking Peptide
Specifications
Specifications
| Gene ID (Entrez) | 2078 |
| Species | Human |
| Product Line | Novus |
| Purity | >50%, by HPLC |
| Content And Storage | Store at -20°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-2472 |
| Gene Alias | transcriptional regulator ERG, 10 ets-related, erg-3, p55, TMPRSS2-ERG prostate cancer specific, transcriptional regulator ERG, Transforming protein ERG, v-ets erythroblastosis virus E26 oncogene homolog (avian), v-ets erythroblastosis virus E26 oncogene like, v-ets erythroblastosis virus E26 oncogene like (avian) |
| Gene Symbol | ERG |
| Quantity | 0.1 mg |
| Research Category | Cancer |
| Show More |