missing translation for 'onlineSavingsMsg'
Learn More

Erbium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta)

Form: Approx. 0.29g/mm, Cast rod, diameter is approximate | CAS: 7440-52-0 | Er | 167.26 g/mol

Supplier:  Thermo Scientific Chemicals 040287DH

 View more versions of this product

Catalog No. AA40287DH


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Erbium
7440-52-0
25 mm
Er
unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic
[Er]
167.26
CHEBI:33379
Silver Gray rod
Silver to Gray
Cast Rod
99.9% (Metals basis excluding Ta)
MFCD00010987
UYAHIZSMUZPPFV-UHFFFAOYSA-N
erbium
23980
Odorless

Chemical Identifiers

7440-52-0
167.26
UYAHIZSMUZPPFV-UHFFFAOYSA-N
23980
erbium
Er
MFCD00010987
unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic
CHEBI:33379
[Er]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P240-P241-P280-P370+P378q

H228

EINECSNumber : 231-160-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only