missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) perchlorate hexahydrate, 50% w/w in aq. soln., Reagent Grade

Catalog No. AA40576NC
Change view
Click to view available options
Quantity:
∼2 g
∼50 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40576NC 10 g
AA40576NA ∼2 g
AA40576NE ∼50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA40576NC Supplier Thermo Scientific Chemicals Supplier No. 040576NC
Only null left

CAS: 14692-15-0 | Cl3ErH12O18 | 573.687 g/mol

Erbium(III) perchlorate hexahydrate is used in nuclear technology for neutron-absorbing control rods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14692-15-0
Molecular Formula Cl3ErH12O18
Molecular Weight (g/mol) 573.687
MDL Number MFCD00150232
InChI Key JZRNHNTYRRYOCR-UHFFFAOYSA-K
Synonym erbium iii perchlorate hexahydrate, erbium perchlorate-water 1/3/6, erbium iii perchlorate hexahydrate w/w in aqueous solution, reagent grade
PubChem CID 20038200
IUPAC Name erbium(3+);triperchlorate;hexahydrate
SMILES O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Er+3]

Specifications

Color Pink
Physical Form Liquid
Quantity 10 g
UN Number UN3211
Solubility Information Micible with water.
Formula Weight 573.70 (465.61 Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w in aq. soln.
Odor Odorless
Chemical Name or Material Erbium(III) perchlorate hexahydrate
Hazard Category H271-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335;
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.
EINECSNumber 237-840-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only