missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) oxalate decahydrate, 99.9%

Catalog No. AAA1842006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1842006 5 g
AAA1842014 25 g
AAA1842022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1842006 Supplier Thermo Scientific Chemicals Supplier No. A1842006
Only null left

CAS: 30618-31-6 | C6H26Er2O22 | 784.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 30618-31-6
Molecular Formula C6H26Er2O22
Molecular Weight (g/mol) 784.77
MDL Number MFCD00150142,MFCD00150142
InChI Key CYHKKMLSGVLNPD-UHFFFAOYSA-N
Synonym erbium oxalate decahydrate,
PubChem CID 131877482
IUPAC Name erbium;oxalic acid;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[Er].[Er].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O

Specifications

Density 2.64
Linear Formula Er2(C2O4)3·10H2O
Quantity 5 g
UN Number UN3288
Sensitivity Hygroscopic
Formula Weight 778.73 (598.58 Anhydrous)
Percent Purity 99.9%
Chemical Name or Material Erbium(III) oxalate decahydrate
Hazard Category H302+H312
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only