missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Erbium(III) chloride hexahydrate
Description
Erbium(III) chloride hexahydrate may be used to prepare upconversion nanocrystal coated silica (UCNP@mSiO2) nanocomposite. The product may be used as a precursor for Er during the doping Y2O3 nanomaterials, which may be useful in bioimaging.
Specifications
Specifications
| CAS | 10025-75-9 |
| Molecular Formula | Cl3Er -+ 6H2O |
| Linear Formula | ErCl3-+6H2O |
| Quantity | 100 g |
| MDL Number | MFCD00149693 |
| Molecular Weight (g/mol) | 381.71 |
| Percent Purity | 99.9% trace metals basis |
| Biological Activity | Respiratory system |